Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568463
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568463 Supplier Invitrogen™ Supplier No. PA568463
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

TSPAN7 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein and may have a role in the control of neurite outgrowth. It is known to complex with integrins. This gene is associated with X-linked mental retardation and neuropsychiatric diseases such as Huntington's chorea, fragile X syndrome and myotonic dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TSPAN7
Gene Accession No. P41732, Q62283
Gene Alias 1200014P11Rik; A15; AI323365; CCG-B7; CD231; CD231 antigen; cell surface glycoprotein A15; DXS1692E; membrane component chromosome X surface marker 1; membrane component, X chromosome, surface marker 1; MRX58; Mxs1; neuroprotective protein 5; PE31; R74651; TALLA; TALLA homolog; TALLA-1; T-cell acute lymphoblastic leukemia associated antigen 1; T-cell acute lymphoblastic leukemia-associated antigen 1; tetraspanin 7; tetraspanin protein; tetraspanin-7; TM4SF2; TM4SF2b; transmembrane 4 superfamily 2b; transmembrane 4 superfamily member 2; Transmembrane 4, superfamily member 2 (cell surface glycoprotein A15); transmembrane protein A15; TSPAN7; tspan-7
Gene Symbols TSPAN7
Host Species Rabbit
Immunogen Synthesized peptide derived from human TSPAN7 [101-150].
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21912, 363447, 7102
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.