Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576938
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576938 Supplier Invitrogen™ Supplier No. PA576938
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

TSPAN7 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein and may have a role in the control of neurite outgrowth. It is known to complex with integrins. This gene is associated with X-linked mental retardation and neuropsychiatric diseases such as Huntington's chorea, fragile X syndrome and myotonic dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene TSPAN7
Gene Accession No. P41732, Q62283
Gene Alias 1200014P11Rik; A15; AI323365; CCG-B7; CD231; CD231 antigen; cell surface glycoprotein A15; DXS1692E; membrane component chromosome X surface marker 1; membrane component, X chromosome, surface marker 1; MRX58; Mxs1; neuroprotective protein 5; PE31; R74651; TALLA; TALLA homolog; TALLA-1; T-cell acute lymphoblastic leukemia associated antigen 1; T-cell acute lymphoblastic leukemia-associated antigen 1; tetraspanin 7; tetraspanin protein; tetraspanin-7; TM4SF2; TM4SF2b; transmembrane 4 superfamily 2b; transmembrane 4 superfamily member 2; Transmembrane 4, superfamily member 2 (cell surface glycoprotein A15); transmembrane protein A15; TSPAN7; tspan-7
Gene Symbols TSPAN7
Host Species Rabbit
Immunogen Recombinant full length Human TSPAN7.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21912, 363447, 7102
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.