Learn More
TSPAN7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 186951AP150UL

Description
The antibody only recognizes TSPAN7 and does not recognize TSPAN6.
TSPAN7 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein and may have a role in the control of neurite outgrowth. It is known to complex with integrins. This gene is associated with X-linked mental retardation and neuropsychiatric diseases such as Huntington′s chorea, fragile X syndrome and myotonic dystrophy.Specifications
TSPAN7 | |
Polyclonal | |
Unconjugated | |
TSPAN7 | |
A15, CCG B7, CD231, Cell surface glycoprotein A15, DXS1692E, Glima, MRX58, MXS1, TALLA 1, tetraspanin 7, TM4SF2, TM4SF2b, Tspan 7, TSPAN7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21912, 7102 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P41732, Q62283 | |
TSPAN7 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.