Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSPAN7/TM4SF2 Antibody (482618), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB9179MFV610
Description
TSPAN7/TM4SF2 Monoclonal antibody specifically detects TSPAN7/TM4SF2 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| TSPAN7/TM4SF2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| metabolism | |
| 7102 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 482618 | |
| Flow Cytometry, CyTOF-ready | |
| A15CCG-B7, CD231, CD231 antigen, Cell surface glycoprotein A15, Membrane component chromosome X surface marker 1, mental retardation, X-linked 58, MRX58, MXS1T-cell acute lymphoblastic leukemia associated antigen 1, T-cell acute lymphoblastic leukemia-associated antigen 1, tetraspanin 7, tetraspanin-7, TM4SF2tetraspanin protein, transmembrane 4 superfamily 2b, Transmembrane 4 superfamily member 2X chromosome, surface marker 1, transmembrane protein A15, Tspan-7 | |
| HEK293 human embryonic kidney cell line transfected with human TSPAN7, Accession # P41732 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction