Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN8 Monoclonal Antibody (657909)
GREENER_CHOICE

Catalog No. PIMA524296
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524296 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524296 Supplier Invitrogen™ Supplier No. MA524296
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This antibody detects mouse TSPAN8 transfectants but not the parental cell line in flow cytometry. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

TSPAN8 (tetraspanin 8), also known as TM4SF3 or CO-029, is a 237 amino acid multi-pass membrane protein that belongs to the tetraspanin family of transmembrane proteins and is thought to play a role in signal transduction events that influence cell development, activation, growth and motility. Localized to the cell membrane, TSPAN8 is expressed in colon, gastric, renal and pancreatic cancers, strongly suggesting a role in cancer formation and metastasis. The gene encoding TSPAN8 maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN8
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 657909
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Tspan8
Gene Accession No. Q8R3G9
Gene Alias C76990; CO-029; CO-29; D6.1A; E330007O21Rik; tetraspanin 8; Tetraspanin8; tetraspanin-8; Tm4sf3; transmembrane 4 superfamily member 3; TSPAN8; Tspan-8; Tumor-associated antigen CO-029
Gene Symbols Tspan8
Host Species Rat
Immunogen CHO Chinese hamster ovary cell line transfected with mouse TSPAN8.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216350
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.