Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599214
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599214 Supplier Invitrogen™ Supplier No. PA599214
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TSPAN8.

TSPAN8 (tetraspanin 8), also known as TM4SF3 or CO-029, is a 237 amino acid multi-pass membrane protein that belongs to the tetraspanin family of transmembrane proteins and is thought to play a role in signal transduction events that influence cell development, activation, growth and motility. Localized to the cell membrane, TSPAN8 is expressed in colon, gastric, renal and pancreatic cancers, strongly suggesting a role in cancer formation and metastasis. The gene encoding TSPAN8 maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Tspan8
Gene Accession No. P19075
Gene Alias C76990; CO-029; CO-29; D6.1A; E330007O21Rik; tetraspanin 8; Tetraspanin8; tetraspanin-8; Tm4sf3; transmembrane 4 superfamily member 3; TSPAN8; Tspan-8; Tumor-associated antigen CO-029
Gene Symbols Tspan8
Host Species Rabbit
Immunogen A synthesized peptide derived from human TSPAN8(Accession P19075), corresponding to amino acid residues F109-A159.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7103
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.