Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSPAN8 Rat anti-Human, BUV496, Clone: 458811, BD Biosciences™
Rat Monoclonal Antibody
Supplier: BD Biosciences 750063
Description
TSPAN8 Monoclonal antibody specifically detects TSPAN8 in Human samples. It is validated for Flow Cytometry
Specifications
TSPAN8 | |
Monoclonal | |
Brilliant Ultraviolet 496 | |
TM4SF3; transmembrane 4 superfamily member 3; tetraspanin-8; tspan-8; CO-029 | |
Human TSPAN8 Transfected Cell Line | |
RUO | |
Human | |
IgG2b κ |
Flow Cytometry | |
458811 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimal conditions that minimize unconjugated dye and antibody. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction