Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN8 Rat anti-Mouse, Brilliant Violet 711, Clone: 657909, BD OptiBuild™

Catalog No. BDB748041 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB748041 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB748041 Supplier BD Biosciences Supplier No. 748041
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TSPAN8 Monoclonal antibody specifically detects TSPAN8 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN8
Applications Flow Cytometry
Classification Monoclonal
Clone 657909
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 711
Host Species Rat
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity TSPAN8
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.