Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSPAN8/TM4SF3 Antibody (657909) [FITC], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB6524F
Description
TSPAN8/TM4SF3 Monoclonal antibody specifically detects TSPAN8/TM4SF3 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
TSPAN8/TM4SF3 | |
Monoclonal | |
FITC | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
657909 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
CO-029, tetraspanin 8, TM4SF3tspan-8, Transmembrane 4 superfamily member 3tetraspanin-8, Tspan-8, Tumor-associated antigen CO-029 | |
CHO Chinese hamster ovary cell line transfected with mouse TSPAN8 | |
0.1 mL | |
Cancer | |
7103 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction