Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPAN9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103149 Supplier Invitrogen™ Supplier No. PA5103149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NET-5.

TSPAN9 includes the tetraspan family protein members which are characterized by four predicted transmembrane domains, and are thought to be involved in physiological processes such as tissue differentiation, immunological responses, and sperm-egg fusion. TSPAN9 has recently been identified as a platelet tetraspanin and a component of tetraspanin microdomains that include the collagen receptor GPVI (glycoprotein VI) and integrin alpha6beta1 but not the von Willebrand receptor GPIbalpha or the integrins alphaIIbbeta3 or alpha2beta1, suggesting that TSPAN9 may act to regulate platelet function in concert with other tetraspanins and their associated proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPAN9
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TSPAN9
Gene Accession No. O75954, Q8BJU2
Gene Alias 6720474K14Rik; 9430079M16Rik; AU018597; NET5; NET-5; new EST tetraspan 5; PP1057; RGD1304740; tetraspan NET-5; tetraspanin 9; tetraspanin-9; transmembrane 4 superfamily member tetraspan NET-5; Tspan9; tspan-9
Gene Symbols TSPAN9
Host Species Rabbit
Immunogen A synthesized peptide derived from human TSPAN9(Accession O75954), corresponding to amino acid residues A143-K193.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10867, 109246
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.