Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPAN9 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989075
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9075 Supplier Bioss Supplier No. BS9448R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSPAN9 includes the tetraspan family protein members which are characterized by four predicted transmembrane domains, and are thought to be involved in physiological processes such as tissue differentiation, immunological responses, and sperm-egg fusion. TSPAN9 has recently been identified as a platelet tetraspanin and a component of tetraspanin microdomains that include the collagen receptor GPVI (glycoprotein VI) and integrin alpha6beta1 but not the von Willebrand receptor GPIbalpha or the integrins alphaIIbbeta3 or alpha2beta1, suggesting that TSPAN9 may act to regulate platelet function in concert with other tetraspanins and their associated proteins.

Specifications

Antigen TSPAN9
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TSPAN9
Gene Accession No. O75954
Gene Alias 6720474K14Rik; 9430079M16Rik; AU018597; NET5; NET-5; new EST tetraspan 5; PP1057; RGD1304740; Tetraspan NET-5; tetraspanin 9; Tetraspanin-9; transmembrane 4 superfamily member tetraspan NET-5; Tspan9; Tspan-9
Gene Symbols TSPAN9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TSPAN9.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10867
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.