Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPEAR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584135 Supplier Invitrogen™ Supplier No. PA584135
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGQEFSVIYK WSHRKLKFTP YQSIATHSAR DWEAFEVDGE HFLAVANHRE GDNHNIDSVI YKWNPATRLF EANQTIATSG AYDWEFFSVG PYSFLVVANT FNGTST Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 88%.

This gene encodes a protein that contains a N-terminal thrombospondin-type laminin G domain and several tandem arranged epilepsy-associated repeats (EARs). A mutation in this gene is the cause of autosomal recessive deafness-98. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Dec 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPEAR
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSPEAR
Gene Accession No. Q8WU66
Gene Alias C21orf29; C330046G03Rik; DFNB98; ORF65; protein TSPEAR; RGD1563108; thrombospondin N-terminal domain/EPTP protein 1; thrombospondin type laminin G domain and EAR repeats; thrombospondin-type laminin G domain and EAR repeat-containing protein; thrombospondin-type laminin G domain and EAR repeats; thrombospondin-type laminin G domain and EAR repeats protein; thrombospondin-type laminin G domain and EAR repeats-containing protein; Tnep1; tspear; TSP-EAR
Gene Symbols TSPEAR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TSPEAR. Recombinant protein control fragment (Product #RP-106432).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54084
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.