Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519088
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519088 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519088 Supplier Invitrogen™ Supplier No. PA519088
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

Mitochondrial peripheral-type benzodiazepine receptor (PBR) is an indispensable element of the steroidogenic machinery, where it mediates the delivery of cholesterol to the inner mitochondrial side chain cleavage cytochrome P-450 upon ligand activation. PBR is composed of three subunits, an isoquinoline binding site, a voltage-dependent anion channel and an adenine nucleotide carrier. PBR is genetically conserved from bacteria to humans and in humans is widely expressed in peripheral organs, whereas in the brain, it is sparse and located mainly in glial cells. Peroxisome proliferator perfluordecanoic acid (PFDA) inhibits the Leydig cell steroidogenesis by affecting PBR mRNA stability, thus inhibiting PBR expression, cholesterol transport into the mitochondria and subsequent steroid formation. A cytoplasmic protein, PRAX-1 ( peripheral benzodiazepine receptor-associated protein 1), is found to specifically interact with PBR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPO
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Tspo
Gene Accession No. P50637
Gene Alias Benzodiazepin receptor (peripheral); benzodiazepine peripheral binding site; benzodiazepine receptor, peripheral; BPBS; BZRP; DBI; IBP; isoquinoline-binding protein; MBR; mDRC; mitochondrial benzodiazepine receptor; OTTHUMP00000028891; PBR; PBS; peripheral-type benzodiazepine receptor; pk18; PKBS; PTBR; Ptbzr; PTBZR02; RATPTBZR02; translocator protein; translocator protein (18kDa); TSPO; Tspo1
Gene Symbols Tspo
Host Species Goat
Immunogen Synthetic peptide sequence (RDNSGRRGGSRLPE) corresponding to the C-terminus amino acids of Tspo (aa 156-169).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12257
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.