Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPP 5% Solution, Reagents

Catalog No. 01333475
Click to view available options
:
Quantity: 20 L

TSPP 5% Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Boiling Point 100°C
Chemical Name or Material TSPP 5% Solution
CAS 7732-18-5, 7722-88-5
Color Colorless
pH >9.9
Physical Form Liquid
Quantity 20 L
Synonym Tetrasodium Pyrophosphate
Percent Purity 5%
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.0312
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.