Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSPY1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534484
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534484 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534484 Supplier Thermo Scientific Supplier No. PA534484
Add to Cart
Add to Cart

TSPY1L Polyclonal Antibody

TSPY1L Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPY1L
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TSPY1
Gene Accession No. Q01534
Gene Alias cancer/testis antigen 78; CT78; DYS14; pJA923; testis specific protein, Y-linked 1; testis-specific Y-encoded protein 1; TSPY; TSPY1; TSPY1L; TSPY1S
Gene Symbols TSPY1
Host Species Rabbit
Immunogen A 13 amino acid peptide near the carboxy terminus of human TSPY1S.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7258
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.