Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSPY3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548840
Description
TSPY3 Polyclonal Antibody for Western Blot
TSPY3 (Testis Specific Protein Y-Linked 3) is a Protein Coding gene. An important paralog of this gene is TSPY4. [GeneCards].
Specifications
TSPY3 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
P0CV98 | |
TSPY3 | |
KLH conjugated synthetic peptide between 277-305 amino acids from the C-terminal region of human TSPY3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TSPY3 | |
Cancer/testis antigen 78; CT78; testis specific protein, Y-linked 3; Testis-specific Y-encoded protein 1; Testis-specific Y-encoded protein 10; Testis-specific Y-encoded protein 3; TSPY; TSPY1; TSPY10; TSPY3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
728137 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction