Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPY3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110530 Supplier Invitrogen™ Supplier No. PA5110530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRPEGSLTYR VPERLRQGFC GVGRAAQALV CASAKEGTAF RMEAVQEGAA GVESEQAALG EEAVLLLDDI MAEVEVVAEE EGLVERREEA QRAQQAVPGP GPMTPESALE ELLAVQVELE PVNAQARKAF SRQREKMERR RKPHLDRRGA VIQSVPGFWA NVIANHPQMS ALITDEDEDM LSYMVSLEVE EEKHPVHLCK IMLFFRSNPY FQNKVITKEY LVNITEYRAS HSTPIEWYPD YEVEAYRRRH HNSSLNFFNW FSDHNFAGSN KIAEILCKDL WRNPLQYYKR MKPPEEGTET SGDSQLLS.

TSPY3 (Testis Specific Protein Y-Linked 3) is a Protein Coding gene. An important paralog of this gene is TSPY4. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPY3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TSPY3
Gene Accession No. P0CV98
Gene Alias Cancer/testis antigen 78; CT78; testis specific protein, Y-linked 3; Testis-specific Y-encoded protein 1; Testis-specific Y-encoded protein 10; Testis-specific Y-encoded protein 3; TSPY; TSPY1; TSPY10; TSPY3
Gene Symbols TSPY3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-308 of human TSPY3 (NP_001071165.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728137
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.