Learn More
Invitrogen™ TSPYL1 Monoclonal Antibody (4F11)

Description
Peptide Sequence: MSGLDGVKRT TPLQTHSIII SDQVPSDQDA HQYLRLRDQS EATQVMAEPG EGGSETVALP PPPPSEEGGV PQDAAGRGGT PQIRVVGGRG HVAIKAGQEE.
Specifications
Specifications
| Antigen | TSPYL1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4F11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TSPYL1 |
| Gene Accession No. | Q9H0U9 |
| Gene Alias | testis-specific protein, Y-encoded-like 1; testis-specific Y-encoded-like protein 1; TSPY like 1; Tspyl; Tspyl1; TSPY-like 1; TSPY-like protein 1; Y encoded |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.