Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPYL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588398
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588398 Supplier Invitrogen™ Supplier No. PA588398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSGLDGVKRT TPLQTHSIII SDQVPSDQDA HQYLRLRDQS EATQVMAEPG EGGSETVALP PPPPSEEGGV PQDAAGRGGT PQIRVVGGRG HVAIKAGQEE GQPPAEGLAA ASVVMAADRS LKKGVQGGEK ALEICGAQRS ASELTAGAEA EAEEVKTGKC ATVSAAVAER ESAEVVKEGL AEKEVMEEQM EVEEQPPEGE EIEVAEEDRL EEEAREEEGP WPLHEALRMD; Positive Samples: U-87MG, SKOV3, LO2, HeLa, 293T, A-549; Cellular Location: Nucleus, nucleolus.

The protein encoded by this gene is found in the nucleolus and is similar to that of a family of genes on the Y-chromosome. This gene is intronless. Defects in this gene are a cause of sudden infant death with dysgenesis of the testes syndrome (SIDDT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPYL1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 4.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TSPYL1
Gene Accession No. Q9H0U9
Gene Alias testis-specific protein, Y-encoded-like 1; testis-specific Y-encoded-like protein 1; TSPY like 1; Tspyl; Tspyl1; TSPY-like 1; TSPY-like protein 1; Y encoded
Gene Symbols TSPYL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human TSPYL1 (NP_0033001).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7259
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.