Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPYL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100796 Supplier Invitrogen™ Supplier No. PA5100796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TSYL2.

This gene encodes the beta-subunit of glucosidase II, an N-linked glycan-processing enzyme in the endoplasmic reticulum (ER). This protein is an acidic phospho-protein known to be a substrate for protein kinase C. Mutations in this gene have been associated with the autosomal dominant polycystic liver disease (PCLD). Alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPYL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TSPYL2
Gene Accession No. Q7TQI8, Q9H2G4
Gene Alias CASK interacting nucleosome assembly protein; CASK-interacting nucleosome assembly protein; CDA1; Cell division autoantigen 1; cell division autoantigen 1 nucleolar protein; CINAP; CTCL; CTCL tumor antigen se20-4; CTCL-associated antigen se20-4; Cutaneous T-cell lymphoma-associated antigen se20-4; cutaneous T-cell lymphoma-associated tumor antigen se20-4; DENTT; differentially expressed nucleolar TGF-beta1 target; differentially-expressed nucleolar TGF-beta1 target protein; DXBwg1396e; DXHXS1008E; E130307F10Rik; HRIHFB2216; NP79; nuclear protein of 79 kDa; nucleolar TGF-beta1 target protein; RGD1563283; RP1-290F12.2; SE204; testis-specific protein Y encoded-like 2; testis-specific Y-encoded-like protein 2; TSPX; TSPY like 2; Tspyl2; TSPY-like 2; TSPY-like protein 2
Gene Symbols TSPYL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TSPYL2(Accession Q9H2G4), corresponding to amino acid residues I452-H502.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 302612, 52808, 64061
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.