Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPYL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111195 Supplier Invitrogen™ Supplier No. PA5111195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NPLRYYLRER GSRIKRKKQE MKKRKTRGRC EVVIMEDAPD YYAVEDIFSE ISDIDETIHD IKISDFMETT DYFETTDNEI TDINENICDS ENPDHNEVPN NETTDNNESA DDHETTDNNE SADDNNENPE DNNKNTDD Highest antigen sequence indentity to the following orthologs: Rat - 54%, Mouse - 55%.

This gene encodes the beta-subunit of glucosidase II, an N-linked glycan-processing enzyme in the endoplasmic reticulum (ER). This protein is an acidic phospho-protein known to be a substrate for protein kinase C. Mutations in this gene have been associated with the autosomal dominant polycystic liver disease (PCLD). Alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPYL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.40 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSPYL2
Gene Accession No. Q9H2G4
Gene Alias CASK interacting nucleosome assembly protein; CASK-interacting nucleosome assembly protein; CDA1; Cell division autoantigen 1; cell division autoantigen 1 nucleolar protein; CINAP; CTCL; CTCL tumor antigen se20-4; CTCL-associated antigen se20-4; Cutaneous T-cell lymphoma-associated antigen se20-4; cutaneous T-cell lymphoma-associated tumor antigen se20-4; DENTT; differentially expressed nucleolar TGF-beta1 target; differentially-expressed nucleolar TGF-beta1 target protein; DXBwg1396e; DXHXS1008E; E130307F10Rik; HRIHFB2216; NP79; nuclear protein of 79 kDa; nucleolar TGF-beta1 target protein; RGD1563283; RP1-290F12.2; SE204; testis-specific protein Y encoded-like 2; testis-specific Y-encoded-like protein 2; TSPX; TSPY like 2; Tspyl2; TSPY-like 2; TSPY-like protein 2
Gene Symbols TSPYL2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TSPYL2. Recombinant protein control fragment (Product #RP-92259).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64061
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.