Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPYL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548329
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548329 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548329 Supplier Invitrogen™ Supplier No. PA548329
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSPYL4 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPYL4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TSPYL4
Gene Accession No. Q8VD63, Q9UJ04
Gene Alias 2610102M01Rik; B230210I21Rik; D10Bwg0791e; dJ486I3.2; KIAA0721; Testis-specific Y-encoded-like protein 4; TSPY like 4; Tspyl4; TSPY-like 4; TSPY-like protein 4
Gene Symbols TSPYL4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 322-350 amino acids from the C-terminal region of human TSPYL4.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23270, 72480
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.