Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPYL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556710 Supplier Invitrogen™ Supplier No. PA556710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQEKEVLSYL NSLEVEELGL ARLGYKIKFY FDRNPYFQNK VLIKEYGCGP SGQVVSRSTP IQWLPGHDLQ SLSQGNPENN RSFFGWFSNH SSIESDKIVE IINEELWPNP LQFYLLSEGA RV Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

Involved in modulation of cell growth and cellular response to gamma radiation probably via regulation of the Akt signaling pathway. Involved in regulation of p53/TP53. Suppresses p53/TP53 protein levels and promotes its ubiquitination; the function is dependent on USP7 and independent on MDM2. Proposed to displace p53/TP53 from interaction with USP7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPYL5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSPYL5
Gene Accession No. Q86VY4
Gene Alias 6330530B20; E130308C19Rik; Kiaa1750; testis-specific protein, Y-encoded-like 5; testis-specific Y-encoded-like protein 5; TSPY like 5; TSPYL5; TSPY-like 5; TSPY-like protein 5
Gene Symbols TSPYL5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TSPYL5. Recombinant protein control fragment (Product #RP-88800).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85453
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.