Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSPYL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144012
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144012 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144012 Supplier Invitrogen™ Supplier No. PA5144012
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human SH-SY5Y whole cell, human Jurkat whole cell, human K562 whole cell, human U20S whole cell. Flow: MCF-7 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Involved in modulation of cell growth and cellular response to gamma radiation probably via regulation of the Akt signaling pathway. Involved in regulation of p53/TP53. Suppresses p53/TP53 protein levels and promotes its ubiquitination; the function is dependent on USP7 and independent on MDM2. Proposed to displace p53/TP53 from interaction with USP7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSPYL5
Applications ELISA, Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene TSPYL5
Gene Accession No. Q86VY4
Gene Alias 6330530B20; E130308C19Rik; Kiaa1750; testis-specific protein, Y-encoded-like 5; testis-specific Y-encoded-like protein 5; TSPY like 5; TSPYL5; TSPY-like 5; TSPY-like protein 5
Gene Symbols TSPYL5
Host Species Rabbit
Immunogen E.coli-derived human TSPYL5 recombinant protein (Position: R24-Q337).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85453
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.