Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531806
Description
Recommended positive controls: A549, H1299, HCT116, MCF-7. Predicted reactivity: Mouse (84%), Rat (85%), Rhesus Monkey (97%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in 20S pre-rRNA processing.
Specifications
TSR2 | |
Polyclonal | |
Unconjugated | |
TSR2 | |
2310007F12Rik; DBA14; DT1P1A10; pre-rRNA-processing protein TSR2 homolog; Tsr2; TSR2 20S rRNA accumulation; TSR2, 20S rRNA accumulation, homolog; TSR2, 20S rRNA accumulation, homolog (S. cerevisiae); TSR2, ribosome maturation factor; WGG motif containing 1; WGG1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90121 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.66 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q969E8 | |
TSR2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 191 of Human TSR2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction