Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556491
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556491 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556491 Supplier Thermo Scientific Supplier No. PA556491
Add to Cart
Add to Cart

TSR2 Polyclonal Antibody

TSR2 Polyclonal Antibody for Western Blot, ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen TSR2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSR2
Gene Accession No. Q969E8
Gene Alias 2310007F12Rik; DBA14; DT1P1A10; pre-rRNA-processing protein TSR2 homolog; Tsr2; TSR2 20S rRNA accumulation; TSR2, 20S rRNA accumulation, homolog; TSR2, 20S rRNA accumulation, homolog (S. cerevisiae); TSR2, ribosome maturation factor; WGG motif containing 1; WGG1
Gene Symbols TSR2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TSR2. Recombinant protein control fragment (Product #RP-92337).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90121
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.