Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598655
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598655 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598655 Supplier Thermo Scientific Supplier No. PA598655
Add to Cart
Add to Cart

TSR2 Polyclonal Antibody

TSR2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TSR2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300, 0.02% sodium azide; pH 7.4
Gene TSR2
Gene Accession No. Q969E8
Gene Alias 2310007F12Rik; DBA14; DT1P1A10; pre-rRNA-processing protein TSR2 homolog; Tsr2; TSR2 20S rRNA accumulation; TSR2, 20S rRNA accumulation, homolog; TSR2, 20S rRNA accumulation, homolog (S. cerevisiae); TSR2, ribosome maturation factor; WGG motif containing 1; WGG1
Gene Symbols TSR2
Host Species Rabbit
Immunogen Recombinant Human Pre-rRNA-processing protein TSR2 homolog protein (1-191AA).
Purification Method Antigen Affinity Chromatography, Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90121
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.