Learn More
TSR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 162631AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0837
Only null left
Add to Cart
Description
Description
May be involved in 20S pre-rRNA processing.
Specifications
Specifications
TSR2
Polyclonal
Unconjugated
TSR2
DT1P1A10, TSR2, WGG1
Rabbit
Antigen Affinity Chromatography
RUO
317418, 69499, 90121
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8C8T8, Q969E8
Tsr2
TSR2 Fusion Protein Ag9328
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.