Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSSC1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297962R
Description
TSSC1 Polyclonal specifically detects TSSC1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TSSC1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
protein TSSC1, tumor suppressing subtransferable candidate 1, Tumor-suppressing STF cDNA 1 protein, Tumor-suppressing subchromosomal transferable fragment candidate gene 1 protein | |
Produced in rabbits immunized with E. coli-derived Human TSSC1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
7260 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction