Learn More
Invitrogen™ TSSC1 Polyclonal Antibody

Description
Immunogen sequence: SKVLTCAAVW RMPKELESGS HESPDDSSST AQTLELLCHL DNTAHGNMAC VVWEPMGDGK KIISLADNHI LLWDLQESSS QAVLASSASL E Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 84%.
Specifications
Specifications
| Antigen | TSSC1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | EIPR1 |
| Gene Accession No. | Q53HC9 |
| Gene Alias | EARP and GARP complex-interacting protein 1; EIPR1; Endosome-associated recycling protein-interacting protein; Golgi-associated retrograde protein-interacting protein; protein TSSC1; Tssc1; tumor suppressing subtransferable candidate 1; tumor-suppressing STF cDNA 1 protein; Tumor-suppressing subchromosomal transferable fragment candidate gene 1 protein |
| Gene Symbols | EIPR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.