Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSSC4 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298057AF350
Description
TSSC4 Polyclonal specifically detects TSSC4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TSSC4 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
protein TSSC4, tumor suppressing subtransferable candidate 4, Tumor-suppressing STF cDNA 4 protein, Tumor-suppressing subchromosomal transferable fragment candidate gene 4 protein | |
Produced in rabbits immunized with E. coli-derived Human TSSC4 fragment. | |
0.1 mL | |
Cancer | |
10078 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction