Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSSC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559737 Supplier Invitrogen™ Supplier No. PA559737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TKYSLEDVTE VSEQSNQATA LAFLGSQSLA APTDCVSSFN QDPSSCGEGR VIFTKPVRGV EARHERKRVL GKVGEPGRGG LGNPATD Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 68%.

TSSC4 is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. This gene is located among several imprinted genes; however, this gene, as well as the pan-hematopoietic expression gene (PHEMX), escapes imprinting. This gene may play a role in malignancies and disease that involve this region. Several transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSSC4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSSC4
Gene Accession No. Q9Y5U2
Gene Alias AA241958; ESTM671070; MNCb-3063; Protein TSSC4; Tssc4; tumor suppressing subtransferable candidate 4; tumor-suppressing STF cDNA 4 protein; tumor-suppressing subchromosomal transferable fragment 4; tumor-suppressing subchromosomal transferable fragment candidate gene 4 protein
Gene Symbols TSSC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TSSC4. Recombinant protein control fragment (Product #RP-97525).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10078
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.