Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSSC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567824
Description
Predicted to react with Mouse and Rat samples.
TSSC4 is one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. This gene is located among several imprinted genes; however, this gene, as well as the pan-hematopoietic expression gene (PHEMX), escapes imprinting. This gene may play a role in malignancies and disease that involve this region. Several transcript variants encoding two different isoforms have been found for this gene.
Specifications
TSSC4 | |
Polyclonal | |
Unconjugated | |
TSSC4 | |
AA241958; ESTM671070; MNCb-3063; Protein TSSC4; Tssc4; tumor suppressing subtransferable candidate 4; tumor-suppressing STF cDNA 4 protein; tumor-suppressing subchromosomal transferable fragment 4; tumor-suppressing subchromosomal transferable fragment candidate gene 4 protein | |
Rabbit | |
affinity chromatography | |
RUO | |
10078, 361682, 56844 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5XIB1, Q9JHE7, Q9Y5U2 | |
TSSC4 | |
Synthesized peptide derived from human TSSC4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction