Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSSK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599844
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599844 Supplier Invitrogen™ Supplier No. PA599844
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TSSK1.

STK22D (TSSK1) is a serine/threonine kinase essential to the function and production of sperm (spermiogenesis). TSSK1 belongs to a family of serine/threonine kinases highly expressed in testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSSK1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TSSK1B
Gene Accession No. Q61241, Q9BXA7
Gene Alias FKSG81; serine/threonine kinase 22A (spermiogenesis associated); serine/threonine kinase 22D (spermiogenesis associated); serine/threonine kinase FKSG81; serine/threonine-protein kinase 22A; spermiogenesis associated; spermiogenesis associated 4; SPOGA1; SPOGA4; Stk22a; STK22D; testis specific serine kinase 1B; testis tissue sperm-binding protein Li 55e; testis-specific kinase 1; testis-specific serine kinase 1; testis-specific serine kinase 1B; testis-specific serine/threonine kinase; testis-specific serine/threonine-protein kinase 1; TSK1; TSK-1; Tssk; TSSK1; TSSK-1; TSSK1B
Gene Symbols Tssk1, TSSK1B
Host Species Rabbit
Immunogen A synthesized peptide derived from human TSSK1B(Accession Q9BXA7), corresponding to amino acid residues P194-E244.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22114, 288358, 83942
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.