Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSSK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570000
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570000 Supplier Invitrogen™ Supplier No. PA570000
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 93%; Guinea Pig: 77%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 86%; Rat: 93%.

STK22B (TSSK2) is a serine/threonine kinase essential for the function and production of sperm (spermiogenesis). Deletions in the STK22B coding region correlate with numerous cardiovascular and glandular malformations. TSSK2 gene ontology annotations related to this gene include intracellular signal transduction; peptidyl-serine phosphorylation; protein autophosphorylation; protein phosphorylation; spermatid development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSSK2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TSSK2
Gene Accession No. Q96PF2
Gene Alias DGSG; DGS-G; DiGeorge syndrome protein G; serine/threonine kinase 22B (spermiogenesis associated); serine/threonine-protein kinase 22B; spermiogenesis associated 2; SPOGA2; STK22B; testicular secretory protein Li 61; testis specific serine kinase 2; testis specific serine threonine kinase 2; testis specific serine/ threonine kinase 2; testis-specific kinase 2; testis-specific serine kinase 2; testis-specific serine/threonine-protein kinase 2; TSK2; TSK-2; TSSK2; TSSK-2
Gene Symbols TSSK2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human TSSK2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23617
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.