Learn More
Invitrogen™ TSSK3 Monoclonal Antibody (6H4)

Description
Peptide Sequence: MEDFLLSNGY QLGKTIGEGT YSKVKEAFSK KHQRKVAIKV IDKMGGPEEF IQRFLPRELQ IVRTLDHKNI IQVYEMLESA DGKICLVMEL AEGGDVFDCV LNGGPLPESR AKALFRQMVE AIRYCHGCGV AHRDLKCENA LLQGFNLKLT DFGFAKVLPK SHRELSQTFC GSTAYAAPEV LQGIPHDSKK GDVWSMGVVL YVMLCASLPF DDTDIPKMLW QQQKGVSFPT HLSISADCQD LLKRLLEPDM ILRPSIEEVS WHPWLAST.
Specifications
Specifications
| Antigen | TSSK3 |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 6H4 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TSSK3 |
| Gene Accession No. | Q96PN8 |
| Gene Alias | 1700014N07Rik; 4930594I21Rik; serine/threonine kinase 22C; serine/threonine kinase 22C (spermiogenesis associated); serine/threonine-protein kinase 22C; spermiogenesis associated; spermiogenesis associated 3; spermiogenesis associated serine/threonine kinase 22D; SPOGA3; STK22C; Stk22d; testis specific serine kinase 3; testis specific serine kinase-3; testis-specific kinase 3; testis-specific serine kinase 3; testis-specific serine/threonine kinase 22C; testis-specific serine/threonine kinase 3b; testis-specific serine/threonine-protein kinase 3; TSK3; TSK-3; Tssk3; Tssk-3; Tssk3b |
| Gene Symbols | TSSK3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.