Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSSK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566343
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566343 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566343 Supplier Invitrogen™ Supplier No. PA566343
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PFDDTNLKKLL RETQKEVTFP ANHTISQECK NLILQMLRQA TKRATILDII KDSWVLKFQP EQPTHEIRLL EAMCQLH Highest antigen sequence identity to the following orthologs - mouse 40%, rat 38%.

TSSK4 belongs to a family of serine/threonine kinases highly expressed in testis (Chen et al., 2005 [PubMed 15964553]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSSK4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSSK4
Gene Accession No. Q6SA08
Gene Alias 1700020B19Rik; 4933424F08Rik; C14orf20; serine/threonine kinase 22E; serine/threonine-protein kinase 22E; STK22E; testis specific serine kinase 4; testis-specific kinase 4; testis-specific serine kinase 4; testis-specific serine/threonine protein kinase 5 variant alpha; testis-specific serine/threonine protein kinase 5 variant beta; testis-specific serine/threonine protein kinase 5 variant delta; testis-specific serine/threonine protein kinase 5 variant gamma; testis-specific serine/threonine-protein kinase 4; TSK4; TSK-4; Tssk4; TSSK-4; TSSK5
Gene Symbols TSSK4
Host Species Rabbit
Immunogen Recombinant Human TSSK4. Recombinant protein control fragment (Product #RP-105156).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283629
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.