Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TSSK4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585021
Description
Immunogen sequence: DFGFAKMVPS NQPVGCSPSY RQVNCFSHLS QTYCGSFAYA CPEILRGLPY NPFLSDTWS.
TSSK4 belongs to a family of serine/threonine kinases highly expressed in testis (Chen et al., 2005 [PubMed 15964553]).
Specifications
TSSK4 | |
Polyclonal | |
Unconjugated | |
TSSK4 | |
1700020B19Rik; 4933424F08Rik; C14orf20; serine/threonine kinase 22E; serine/threonine-protein kinase 22E; STK22E; testis specific serine kinase 4; testis-specific kinase 4; testis-specific serine kinase 4; testis-specific serine/threonine protein kinase 5 variant alpha; testis-specific serine/threonine protein kinase 5 variant beta; testis-specific serine/threonine protein kinase 5 variant delta; testis-specific serine/threonine protein kinase 5 variant gamma; testis-specific serine/threonine-protein kinase 4; TSK4; TSK-4; Tssk4; TSSK-4; TSSK5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
283629 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6SA08 | |
TSSK4 | |
Recombinant protein corresponding to Human TSSK4. Recombinant protein control fragment (Product #RP-108288). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction