Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSSK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596837 Supplier Invitrogen™ Supplier No. PA596837
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLRHKYLINF YRAIESTSRV YIILELAQGG DVLEWIQRYG ACSEPLAGKW FSQLTLGIAY LHSKSIVHRD LKLENLLLDK WENVKISDFG FAKMVPSNQP VGCSPSYRQV NCFSHLSQTY CGSFAYACPE ILRGLPYNPF LSDTWSMGVI LYTLVVAHLP FDDTNLKKLL RETQKEVTFP ANHTISQECK NLILQMLRQA TKRATILDII KDSWVLKFQP EQPTHEIRLL EAMCQLHNTT KQHQSLQITT; Positive Samples: Mouse testis.

TSSK4 belongs to a family of serine/threonine kinases highly expressed in testis (Chen et al., 2005 [PubMed 15964553]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSSK4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TSSK4
Gene Accession No. Q9D411
Gene Alias 1700020B19Rik; 4933424F08Rik; C14orf20; serine/threonine kinase 22E; serine/threonine-protein kinase 22E; STK22E; testis specific serine kinase 4; testis-specific kinase 4; testis-specific serine kinase 4; testis-specific serine/threonine protein kinase 5 variant alpha; testis-specific serine/threonine protein kinase 5 variant beta; testis-specific serine/threonine protein kinase 5 variant delta; testis-specific serine/threonine protein kinase 5 variant gamma; testis-specific serine/threonine-protein kinase 4; TSK4; TSK-4; Tssk4; TSSK-4; TSSK5
Gene Symbols TSSK4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 79-328 of human TSSK4 (NP_7776042).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71099
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.