Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSSK6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571708
Description
The TSSK6 gene is an intronless gene encodes a member of the CAMK (calcium/calmodulin-dependent) serine/threonine protein kinase family. The encoded kinase has a broad expression pattern but is described as testis-specific due to effects on fertility. Male mice which lack the gene encoding a highly similar protein are sterile and have morphologically abnormal sperm. [provided by RefSeq, Jan 2012]
Specifications
TSSK6 | |
Polyclonal | |
Unconjugated | |
TSSK6 | |
Testis-specific serine/threonine-protein kinase 6; TSK-6; TSSK-6; Testis-specific kinase 6; Cancer/testis antigen 72; CT72; Serine/threonine-protein kinase SSTK; Small serine/threonine kinase; TSSK6 {ECO:0000312/EMBL:AAH146111} | |
Rabbit | |
selective precipitation | |
RUO | |
83983 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q9BXA6 | |
TSSK6 | |
KLH conjugated synthetic peptide between 25-54 amino acids from the N-terminal region of human TSSK6 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction