Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TST Mouse anti-Human, Mouse, Rat, Clone: 2A6C5, Proteintech
SDP

Catalog No. 501736461 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6461 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6461 Supplier Proteintech Group Inc Supplier No. 660181IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The product of this gene is a mitochondrial matrix enzyme that is encoded by the nucleus. It may play roles in cyanide detoxification, the formation of iron-sulfur proteins, and the modification of sulfur-containing enzymes. The gene product contains two highly conservative domains (rhodanese homology domains), suggesting these domains have a common evolutionary origin.

Specifications

Antigen TST
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2A6C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TST
Gene Accession No. P24329, P52196, Q16762
Gene Alias RDS, Rhodanese, Thiosulfate sulfurtransferase, TST
Gene Symbols TST
Host Species Mouse
Immunogen TST Fusion Protein Ag9235
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22117, 25274, 7263
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.