Learn More
Invitrogen™ TSTA3 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (92%), Rat (92%), Chicken (82%), Rhesus Monkey (98%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | TSTA3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | TSTA3 |
| Gene Accession No. | Q13630, Q8K3X2 |
| Gene Alias | 3-5 epimerase/4-reductase; AI256181; FX; GDP-4-keto-6-deoxy-D-mannose epimerase-reductase; GDP-4-keto-6-deoxy-D-mannose-3,5-epimerase-4-reductase; GDP-fucose synthase; GDP-L-fucose synthase; GDP-L-fucose synthetase; GFUS; LOC513158 protein; P35B; Protein FX; red cell NADP(H)-binding protein; SDR4E1; short chain dehydrogenase/reductase family 4E, member 1; Short-chain dehydrogenase/reductase family 4E member 1; testis tissue sperm-binding protein Li 45a; tissue specific transplantation antigen 3; tissue specific transplantation antigen P35B; transplantation antigen P35B; Tsta3; Tstap35b; Tum-P35B antigen |
| Gene Symbols | TSTA3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.