Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSTD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552316
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552316 Supplier Invitrogen™ Supplier No. PA552316
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGAPTVSLP ELRSLLASGR ARLFDVRSRE EAAAGTIPGA LNIPVSELES ALQMEPAAFQ ALYSAEKPKL EDEHLVFFCQ MGKRGLQATQ LARSLGYTGY G Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.

Thiosulfate:glutathione sulfurtransferase (TST) required to produce S-sulfanylglutathione (GSS(-)), a central intermediate in hydrogen sulfide metabolism (PubMed:24981631). Provides the link between the first step in mammalian H(2)S metabolism performed by the sulfide:quinone oxidoreductase (SQOR) which catalyzes the conversion of H(2)S to thiosulfate, and the sulfur dioxygenase (SDO) which uses GSS(-) as substrate (PubMed:24981631). The thermodynamic coupling of the irreversible SDO and reversible TST reactions provides a model for the physiologically relevant reaction with thiosulfate as the sulfane donor (PubMed:24981631). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSTD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSTD1
Gene Accession No. Q8NFU3
Gene Alias KAT; putative thiosulfate sulfurtransferase KAT; thiosulfate sulfurtransferase (rhodanese)-like domain containing 1; thiosulfate sulfurtransferase KAT, putative; thiosulfate sulfurtransferase like domain containing 1; thiosulfate sulfurtransferase/rhodanese-like domain-containing protein 1; Thiosulfate:glutathione sulfurtransferase; TST; TSTD1
Gene Symbols TSTD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TSTD1. Recombinant protein control fragment (Product #RP-89177).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131187
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.