Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TSTD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555024 Supplier Invitrogen™ Supplier No. PA555024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPGIHLSPGE FHKEVEKFLS QANQEQSDTI LLDCRNFYES KIGRFQGCLA PDIRKFSYFP SYVDKNLELF REKRVLMYCT GGIRCERGSA YLKAKGVC Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 91%.

TSTD2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSTD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TSTD2
Gene Accession No. Q5T7W7
Gene Alias C9orf97; PP4189; rhodanese domain-containing protein 2; rhodanese like domain containing 2; thiosulfate sulfurtransferase (rhodanese)-like domain containing 2; thiosulfate sulfurtransferase like domain containing 2; thiosulfate sulfurtransferase -like domain containing 2; thiosulfate sulfurtransferase/rhodanese-like domain-containing protein 2; TSTD2
Gene Symbols TSTD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TSTD2. Recombinant protein control fragment (Product #RP-92973).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158427
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.