Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSV Value Undercounter Refrigerator

Catalog No. p-9255004
Change view
Click to view available options
Type:
General Purpose ADA Refrigerator
General Purpose Refrigerator
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
TSV05RPAA General Purpose ADA Refrigerator
TSV05RPSA General Purpose Refrigerator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. TSV05RPAA Supplier Thermo Scientific™ Supplier No. TSV05RPAA
Only null left
Add to Cart
Add to Cart
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Chill, freeze and retrieve samples safely, simply and economically with the Thermo Scientific™ TSV Value™ Undercounter Refrigerator

Chill, freeze and retreive samples safely, simply and economically with the Thermo Scientific™ TSV Value™ Undercounter Refrigerator

  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally friendly, SNAP compliant hydrocarbon refrigerants and foam
  • Adjustable temperature control
  • Easy to clearn, durable finish
TRUSTED_SUSTAINABILITY

Specifications

Type General Purpose ADA Refrigerator
Capacity (English) 5.5 cu. ft.
Capacity (Metric) 155.7 L
Temperature Range 1°C to 12°C
Temperature (Setpoint) Adjustable
Refrigerant R600a hydrocarbon refrigerant
Door Style Hinged
No. of Doors 1
Hinge Side Right
Shelves 3 Adjustable
Exterior Finish Enameled Steel
Insulation Cyclopentane
Interior ABS Plastic
Certifications/Compliance cULus
Casters 2 adjustable feet in front
Color White
Control Type Mechanical Dial
For Use With (Application) Indoor General Storage of Non-Critical Samples and Reagents
Typical Lifetime 10 Years
Warranty 12 Months
Defrost Manual
Min. Clearance 4 in. on all sides
Regulatory Approvals ADA Compliant
Access Security Key Lock
Compressor Hermetic
Dimensions (DxWxH) Exterior 23.5 x 23.63 x 32.5 in. (59.69 x 60.02 x 82.6cm)
Dimensions (D x W x H) Interior 17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5 cm)
Amperage 0.8 A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115V 60Hz
Frequency 60 Hz
Energy Usage .31 kWh/Day
Shipping Weight (English) 95 lb.
Shipping Weight (Metric) 43 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.