Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTBK1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306356PCP
Description
TTBK1 Polyclonal antibody specifically detects TTBK1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
TTBK1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
BDTK, Brain-derived tau kinase, EC 2.7.11, EC 2.7.11.1, KIAA1855tau-tubulin kinase 1, RP3-330M21.4, tau tubulin kinase 1 | |
Produced in rabbits immunized with E. coli-derived Human TTBK1 fragment. | |
0.1 mL | |
Protein Kinase | |
84630 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction