Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTBK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556840
Description
Immunogen sequence: RLVMEKRQGR LLLRLASGAS SSSSEEQRRA SETLSGTGSE EDTPASEPAA ALPRKSGRAA ATRSRIPRPI GLRMPMPVAA QQ Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.
TTBK1 is a member of the casein kinase 1 superfamily, and it functions as a neuron-specific serine/threonine and tyrosine kinase. Its primary role is to regulate the phosphorylation of tau, a protein that stabilizes microtubule assemblies. It has been found that genetic variants in this gene are linked to Alzheimer's disease. Additionally, TTBK1 is associated with several other diseases, including Childhood-Onset Schizophrenia and Spinocerebellar Ataxia 11.
Specifications
TTBK1 | |
Polyclonal | |
Unconjugated | |
TTBK1 | |
AU017937; AW048023; BDTK; Brain-derived tau kinase; C330008L01Rik; KIAA1855; RP3-330M21.4; tau tubulin kinase 1; tau-tubulin kinase 1; Ttbk1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84630 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TCY1 | |
TTBK1 | |
Recombinant protein corresponding to Human TTBK1. Recombinant protein control fragment (Product #RP-94885). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction