Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTBK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583207 Supplier Invitrogen™ Supplier No. PA583207
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPPQLSQGDG RSETSQPPTP GSPSHSPLHS GPRPRRRESD PTGPQRQVFS VAPPFEVNGL PRAVPLSLPY QDFKRDLSDY RERARLLNRV R Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 97%.

TTBK1 is a member of the casein kinase 1 superfamily, and it functions as a neuron-specific serine/threonine and tyrosine kinase. Its primary role is to regulate the phosphorylation of tau, a protein that stabilizes microtubule assemblies. It has been found that genetic variants in this gene are linked to Alzheimer's disease. Additionally, TTBK1 is associated with several other diseases, including Childhood-Onset Schizophrenia and Spinocerebellar Ataxia 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTBK1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTBK1
Gene Accession No. Q5TCY1
Gene Alias AU017937; AW048023; BDTK; Brain-derived tau kinase; C330008L01Rik; KIAA1855; RP3-330M21.4; tau tubulin kinase 1; tau-tubulin kinase 1; Ttbk1
Gene Symbols TTBK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTBK1. Recombinant protein control fragment (Product #RP-105863).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84630
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.