Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTBK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553728 Supplier Invitrogen™ Supplier No. PA553728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTEPLDVTKT QTFSVVPNQD KNNEIMKLLT VGTSEISSRD IDPHVEGQIG QVAEMQKNKI SKDDDIMSED LPGHQGDLST FLHQEGKREK ITPRNGELFH CVS Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 78%.

Tau tubulin kinase (TTBK2) is a serine/threonine kinase whose substrates are thought to include the tau and tubulin proteins. Mutations in the TTBK2 gene cause spinocerebellar ataxia type 11 (SCA11), a neurodegenerative disease characterized by progressive ataxia and atrophy of the cerebellum and brainstem. Recent studies have shown that increased levels of TTBK2 expression in kidney carcinoma and melanoma cell lines increases resistance to the chemotherapeutic agent Sunitinib, and that reduction of TTBK2 though the use of siRNA sensitized these cell lines to the effects of Sunitib, suggesting that TTBK2 can be used as a resistance marker as well a potential target to overcome resistance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTBK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTBK2
Gene Accession No. Q6IQ55
Gene Alias 2610507N02Rik; AI326283; B930008N24Rik; Bby; KIAA0847; mKIAA0847; Protein bartleby; SCA11; tau tubulin kinase 1; tau tubulin kinase 2; tau-tubulin kinase 2; Ttbk; Ttbk1; TTBK2; TTK
Gene Symbols TTBK2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTBK2. Recombinant protein control fragment (Product #RP-91429).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146057
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.