Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557715
Description
Immunogen sequence: PSIHQAREAC MRLPKQIEER NERLKEEMLG KLKDLGNLVL RPFGLSTENF QIKQDSSTGS YSINFVQNPN NNR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.
This gene encodes a protein that belongs to the tetratrico peptide repeat superfamily of proteins. The encoded protein plays a role in protein-protein interactions, and binds to the Galpha subunit of G protein-coupled receptors to activate the Ras signaling pathway. Alternate splicing results in multiple transcript variants.
Specifications
TTC1 | |
Polyclonal | |
Unconjugated | |
Ttc1 | |
4833412C19Rik; C79328; tetratricopeptide repeat domain 1; tetratricopeptide repeat protein 1; TPR repeat protein 1; TPR1; Ttc1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7265 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99614 | |
Ttc1 | |
Recombinant protein corresponding to Human TTC1. Recombinant protein control fragment (Product #RP-96823). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction